MilliporeSigma
  • Morphology-controlled flame synthesis of single, branched, and flower-like α-MoO3 nanobelt arrays.
Morphology-controlled flame synthesis of single, branched, and flower-like α-MoO3 nanobelt arrays.
Nano letters (2011-01-26)
Lili Cai, Pratap M Rao, Xiaolin Zheng
ABSTRACT
We report an atmospheric, catalyst-free, rapid flame synthesis technique for growing single, branched, and flower-like α-MoO(3) nanobelt arrays on diverse substrates. The growth rate, morphology, and surface coverage density of the α-MoO(3) nanobelts were controlled by varying the flame equivalence ratio, the source temperature, the growth substrate temperature, and the material and morphology of the growth substrate. This flame synthesis technique is a promising, alternative way to synthesize one-dimensional metal oxide nanostructures in general.
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%