MilliporeSigma
  • Determination of the epimerization rate constant of amygdalin by microemulsion electrokinetic chromatography.
Determination of the epimerization rate constant of amygdalin by microemulsion electrokinetic chromatography.
Electrophoresis (2011-01-22)
Lishuang Yu, Hongzhi Ye, Lili Zheng, Lidian Chen, Kedan Chu, Xianxiang Liu, Xueqin Xu, Guonan Chen
ABSTRACT
A new method for separation and determination of amygdalin and its epimer (neoamygdalin) in the epimerization of amygdalin by MEEKC is proposed. For the chiral separation of amygdalin and neoamygdalin, a running buffer composed of 80 mM sodium cholate, 5.0% v/v butan-1-ol, 0.5% v/v heptane and 94.5% v/v 30 mM Na(2) B(4) O(7) buffer (pH 9.00) is proposed. Under optimum conditions, the basic separation of amygdalin and neoamygdalin can be achieved within 7 min. The calibration curve for amygdalin showed excellent linearity in the concentration range of 20-1000 μg/mL with a detection limit of 5.0 μg/mL (S/N=3). The epimerization rate constant of amygdalin in basic microemulsion was first determined by monitoring the concentration changes of amygdalin, and the epimerization rate constant of amygdalin was found to be 2×10(-3) min(-1) at 25°C under the above optimum microemulsion conditions.
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels