MilliporeSigma
  • Inhibition of CD44 N- and O-linked glycosylation decreases endometrial cell lines attachment to peritoneal mesothelial cells.
Inhibition of CD44 N- and O-linked glycosylation decreases endometrial cell lines attachment to peritoneal mesothelial cells.
Fertility and sterility (2010-10-26)
Allison K Rodgers, Anitha Nair, Peter A Binkley, Rajeshwar Tekmal, Robert S Schenken
ABSTRACT
The attachment of endometrial epithelial cells (EECs) and endometrial stromal cells (ESCs) to peritoneal mesothelial cells (PMCs) with and without inhibition of N- and O-linked glycosylation, the viability of EECs and ESCs, and the expression of CD44 surface density were evaluated. Inhibition of CD44 N- and O-linked glycosylation by using tunicamycin and/or B-GalNAc statistically significantly inhibited endometrial cell attachment to peritoneal mesothelial cells, suggesting a role in establishment of early endometriotic lesions.
Sigma-Aldrich
Benzyl 2-acetamido-2-deoxy-α-D-galactopyranoside, O-glycosylation inhibitor, ≥97% (TLC)