- A new strategy of photoelectrochemical analysis without an external light source based on isoluminol chemiluminescence probe.
A new strategy of photoelectrochemical analysis without an external light source based on isoluminol chemiluminescence probe.
Chemical communications (Cambridge, England) (2010-09-25)
Caifeng Ding, Hui Li, Xiangling Li, Shusheng Zhang
PMID20865188
ABSTRACT
A new strategy of photoelectrochemical (PEC) analysis with chemiluminescence (CL) of the isoluminol-H(2)O(2)-Co(2+) system as a light source for the determination of physiological thiols in cancer cells was reported for the first time.
MATERIALS