MilliporeSigma
A new bianthracene C-arabinopyranoside from Senna septemtrionalis.
Natural product communications (2010-06-05)
Gizachew Alemayehu, Legesse Adane, Berhanu M Abegaz
ABSTRACT
Chrysophanol, physcion, emodin, floribundone-1, 5,7'-physcion-fallacinol, and the novel 5,7'-physcion-physcion-10'-C-alpha-arabinopyranoside were isolated from the stem bark of Senna septemtrionalis. The structures of these secondary metabolites were determined on the basis of spectroscopic analysis, especially from NMR spectra in conjunction with COSY, HMQC, HMBC and TOCSY.
Product Description
Supelco
Physcion, analytical standard
Sigma-Aldrich
Physcion, ≥98.0% (TLC)