MilliporeSigma
  • Supplemental selenium alleviates the toxic effects of excessive iodine on thyroid.
Supplemental selenium alleviates the toxic effects of excessive iodine on thyroid.
Biological trace element research (2010-06-03)
Jian Xu, Xiao-Li Liu, Xue-Feng Yang, Huai-Lan Guo, Li-Na Zhao, Xiu-Fa Sun
ABSTRACT
As excessive iodine intake is associated with a decrease of the activities of selenocysteine-containing enzymes, supplemental selenium was hypothesized to alleviate the toxic effects of excessive iodine. In order to verify this hypothesis, Balb/C mice were tested by giving tap water with or without potassium iodate and/or sodium selenite for 16 weeks, and the levels of iodine in urine and thyroid, the hepatic selenium level, the activities of glutathione peroxidase (GSHPx), type 1 deiodinase (D1), and thyroid peroxidase (TPO) were assayed. It had been observed in excessive iodine group that hepatic selenium, the activities of GSHPx, D1, and TPO decreased, while in the groups of 0.2 mg/L, 0.3 mg/L and 0.4 mg/L supplemental selenium, the urinary iodine increased significantly. Compared with the group of excessive iodine intake alone, supplemental selenium groups had higher activities of GSHPx, D1, and TPO. We could draw the conclusion that supplemental selenium could alleviate toxic effect of excessive iodine on thyroid. The optimal dosage of selenium ranges from 0.2 to 0.3 mg/L which can protect against thyroid hormone dysfunction induced by excessive iodine intake.
Sigma-Aldrich
Potassium iodate solution, 1/60 M
Sigma-Aldrich
Potassium iodate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium iodate, 99.995% trace metals basis
Supelco
Potassium iodate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium iodate, reagent grade, ≥98%
Sigma-Aldrich
Potassium iodate, ACS reagent, 99.5%