MilliporeSigma
Microarray analyses of shrimp immune responses.
Marine biotechnology (New York, N.Y.) (2010-04-16)
Takashi Aoki, Han-Ching Wang, Sasimanas Unajak, Mudjekeewis D Santos, Hidehiro Kondo, Ikuo Hirono
ABSTRACT
Shrimp aquaculture is one of the major foodproducing industries in the world. However, it is being impacted by several problems including diseases, antibiotic use, and environmental factors. The extent of the effects of these problems in the immune system of the shrimp at the molecular level is just beginning to be understood. Here, we review the gene expression profile of shrimp in response to some of these problems using the high-throughput microarray analysis, including white spot syndrome virus, yellow head virus, Vibrio spp., peptidoglycan, oxytetracycline, oxolinic acid, salinity, and temperature.
Sigma-Aldrich
Oxolinic acid, quinolone antibiotic
Supelco
Oxolinic acid, analytical standard