MilliporeSigma
  • Assessment of cytocompatibility of surface-modified CdSe/ZnSe quantum dots for BALB/3T3 fibroblast cells.
Assessment of cytocompatibility of surface-modified CdSe/ZnSe quantum dots for BALB/3T3 fibroblast cells.
Toxicology in vitro : an international journal published in association with BIBRA (2010-04-07)
Sanjeev Kumar Mahto, Chansik Park, Tae Hyun Yoon, Seog Woo Rhee
ABSTRACT
With the widespread use of quantum dots (QDs), the likelihood of exposure to QDs has been assumed to have increased substantially. Recently, QDs have been employed in numerous biological and medical applications. However, there is a lack of toxicological data pertaining to QDs. In this study, we aimed to investigate the cytocompatibility of surface-modified CdSe/ZnSe QDs for BALB/3T3 fibroblast cells. The ligands used for surface modification are mercaptopropionic acid (MPA) and Gum arabic (GA)/tri-n-octylphosphine oxide (TOPO). Cells were exposed to different concentrations of QDs followed by illustrative cytotoxicity analyses. Furthermore, we used a confocal microscope to assess intracellular uptake of QDs. Confocal images showed that MPA-coated QDs were distributed inside the cytoplasmic region of cells. In contrast, GA/TOPO-coated QDs were not found inside cells. MPA-coated QDs were highly cytocompatible, whereas GA/TOPO-coated QDs were toxic to the cells. Cells treated with GA/TOPO-coated QDs showed altered morphology, decreased viability, significant concentrations of intracellular free cadmium, detectable reactive oxygen species (ROS) formation, depolymerized cytoskeleton, and irregular-shaped nuclei. This study suggests that surface modification by ligands plays a significant role in the prevention of cytotoxicity of QDs.
Sigma-Aldrich
Trioctylphosphine oxide, for extraction analysis, ≥98.5%
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%