MilliporeSigma
  • Biocatalytic properties of a recombinant Fusarium proliferatum lactonase with significantly enhanced production by optimal expression in Escherichia coli.
Biocatalytic properties of a recombinant Fusarium proliferatum lactonase with significantly enhanced production by optimal expression in Escherichia coli.
Applied biochemistry and biotechnology (2009-10-31)
Bing Chen, Li-Qiang Fan, Jian-He Xu, Jian Zhao, Xian Zhang, Li-Ming Ouyang
ABSTRACT
The levo-lactonase gene of Fusarium proliferatum ECU2002 (EC3.1.1.25) was cloned and expressed in Escherichia coli JM109 (DE3) for biocatalytic resolution of industrially important chiral lactones, including DL-pantoyl lactone which was a key precursor to calcium D-pantothenate. By increasing the biomass concentration and lowering the inducer (isopropyl-beta-D-thiogalactoside) concentration and induction temperature, the lactonase production was significantly enhanced up to 20 kU/L, which was 20 times higher than that of wild-type strain F. proliferatum ECU2002. The recombinant Fusarium lactonase was purified using immobilized metal affinity chromatography, and its SDS-PAGE revealed a molecular mass of 50 kDa for the recombinant protein, suggesting that the enzyme was a simplex protein. Furthermore, biocatalytic properties of the recombinant lactonase were investigated, including kinetic parameters, additive's effect, and substrate specificity. The results reported in this paper provide a feasible method to make the whole cells of E. coli JM109 (DE3) expressing lactonase gene to be a highly efficient and easy-to-make biocatalyst for asymmetric synthesis of chiral compounds.
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%