MilliporeSigma
  • Assessment of dermal absorption by thermogravimetric analysis: Development of a diffusion model based on Fick's second law.
Assessment of dermal absorption by thermogravimetric analysis: Development of a diffusion model based on Fick's second law.
Journal of pharmaceutical sciences (2009-03-14)
Matias Rauma, Gunnar Johanson
ABSTRACT
Recent work in our laboratory shows that it is possible to reproducibly measure the weight change over time of a piece of porcine skin exposed to a chemical vapor by thermogravimetric analysis (TGA), that is, using a microbalance maintained at constant temperature and humidity. Here, we develop a diffusion model by which the TGA skin weight curve can be interpreted. A TGA-specific analytical solution for Fick's second law of diffusion was derived. The solution contains two chemical-dependent parameters; a diffusion coefficient (D) and a skin/air partition coefficient (P). The resulting function was fitted to weight curves from 83 experiments with 4 chemicals; n-butyl acetate, methanol, 2-propanol, and toluene. In most experiments, a single weight function could not adequately describe the shape of the weight curve, therefore up to five parallel compartments, each with its parameter set Di and Pi, were tested. A two-compartment model was adequate for 58% of the experiments according to the Akaike Information Criterion. Dermal penetration of the chemicals was also measured with Franz diffusion cells. The diffusion coefficients obtained by the two methods agreed well.
Sigma-Aldrich
Butyl acetate, JIS special grade, ≥99.0%
Sigma-Aldrich
Butyl acetate, ≥99.5%, suitable for atomic absorption spectrometry
Sigma-Aldrich
Butyl acetate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Supelco
Butyl acetate, analytical standard
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%