MilliporeSigma
  • Response of the egg parasitoids Trissolcus basalis and Telenomus podisi to compounds from defensive secretions of stink bugs.
Response of the egg parasitoids Trissolcus basalis and Telenomus podisi to compounds from defensive secretions of stink bugs.
Journal of chemical ecology (2009-01-08)
Raúl A Laumann, Michely F S Aquino, Maria C B Moraes, Martín Pareja, Miguel Borges
ABSTRACT
We tested the hypotheses that host-searching behavior of the egg parasitoids Telenomus podisi and Trissolcus basalis may be differentially influenced by the different blends of volatiles released from the metathoracic glands of adult stink bug host species. We further studied whether such a differential response is due to different individual components of these glands and whether these responses reflect host preferences. Y-tube olfactometer bioassays were carried out with crude extracts of metathoracic glands of five different host species of neotropical stink bugs. Additionally, we tested the parasitoids' responses to synthetic standards of individual compounds identified in these stink bug glands. Results showed that females of T. basalis and T. podisi responded differentially to crude gland extracts of the different species of host stink bugs and to the compounds tested. The parasitoid T. basalis showed a positive taxic behavior to Nezara viridula methathoracxic gland extracts of a host species preferred in the field, i.e., N. viridula. Furthermore, T. basalis responded positively to 4-oxo-(E)-2-hexenal and (E)-2-decenal, two components of N. viridula glandular secretion. Higher residence time, reduced linear velocity, and higher tortuosity in the arm of the olfactometer supplied with 4-oxo-(E)-2-hexenal showed that this compound modifies the kinetics of some traits of T. basalis walking pattern and suggests that it might stimulate the searching behavior of this parasitoid. The parasitoid T. podisi was attracted to crude gland extracts of the preferred host (Euschistus heros) and also to 4-oxo-(E)-2-hexenal. Additionally, this parasitoid responded positively to (E)-2-hexenal and to the hydrocarbon tridecane, both of which are defensive compounds released from the metathoracic glands by several stink bugs. The results indicate some degree of specialization in the response of two generalist parasitoid species toward defensive secretions of stink bugs.
Product Description
Supelco
Tridecane, analytical standard
Sigma-Aldrich
Tridecane, ≥99%