MilliporeSigma
  • The role of short-range diffusion in solvent-assisted mechanochemical synthesis of metal complexes.
The role of short-range diffusion in solvent-assisted mechanochemical synthesis of metal complexes.
Dalton transactions (Cambridge, England : 2003) (2008-10-02)
Graham A Bowmaker, John V Hanna, Robert D Hart, Brian W Skelton, Allan H White
ABSTRACT
The role of short-range diffusion in solvent-assisted mechanochemical synthesis is demonstrated in studies of a polymorphic transition and a ligand dissociation reaction involving copper(I) thiocyanate complexes.
Sigma-Aldrich
Copper(I) thiocyanate, 99%