MilliporeSigma
  • Reduction of lead oxide (PbO2) by iodide and formation of iodoform in the PbO2/I(-)/NOM system.
Reduction of lead oxide (PbO2) by iodide and formation of iodoform in the PbO2/I(-)/NOM system.
Environmental science & technology (2008-05-24)
Yi-Pin Lin, Michael P Washburn, Richard L Valentine
ABSTRACT
Lead oxide (PbO2) can be an important form of lead mineral scale occurring in some water distribution systems. It is believed to be formed by the oxidation of lead-containing plumbing materials by free chlorine. Its reactivity in water, however, has not been well studied. Iodide is also found in source drinking waters, albeit at low concentrations. Consideration of thermodynamics suggests that iodide can be oxidized by PbO2. In this investigation, iodide ion was used as a probe compound to study the reduction of PbO2 and the formation of iodoform, which has been predicted to be a carcinogen, in the presence of natural organic matter (NOM). The reduction of PbO2 by iodide can be expressed as PbO2 + 31(-) + 4H+ --> Pb(2+) + I3(-) + 2H2O, and the reaction kinetics has been determined in this study. In the presence of NOM, I3- reacts with NOM to form iodoform and its concentration is proportional to the NOM concentration. Our results indicate that PbO2 is a very powerful oxidant and can possibly serve as an oxidant reservoir for the formation of iodinated disinfection byproduct through a novel reaction pathway.
Sigma-Aldrich
Iodoform, SAJ first grade
Sigma-Aldrich
Iodoform, 99%