MilliporeSigma
  • Melanin-based high-throughput screen for L-tyrosine production in Escherichia coli.
Melanin-based high-throughput screen for L-tyrosine production in Escherichia coli.
Applied and environmental microbiology (2007-12-25)
Christine Nicole S Santos, Gregory Stephanopoulos
ABSTRACT
We present the development of a simple, high-throughput screen for identifying bacterial strains capable of L-tyrosine production. Through the introduction of a heterologous gene encoding a tyrosinase, we were able to link L-tyrosine production in Escherichia coli with the synthesis of the black and diffusible pigment melanin. Although melanin was initially produced only at low levels in morpholinepropanesulfonic acid (MOPS) minimal medium, phosphate supplementation was found to be sufficient for increasing both the rates of synthesis and the final titers of melanin. Furthermore, a strong linear correlation between extracellular L-tyrosine content and melanin formation was observed by use of this new medium formulation. A selection strategy that utilizes these findings has been developed and has been shown to be effective in screening large combinatorial libraries in the search for L-tyrosine-overproducing strains.
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)