MilliporeSigma
Fragrance material review on ionone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
J Lalko, A Lapczynski, D McGinty, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of ionone when used as a fragrance ingredient is presented.
Sigma-Aldrich
α-Ionone, ≥90%, stabilized
Sigma-Aldrich
α-Ionone, technical grade, 90%