MilliporeSigma
  • Synthesis of hyaluronan haloacetates and biology of novel cross-linker-free synthetic extracellular matrix hydrogels.
Synthesis of hyaluronan haloacetates and biology of novel cross-linker-free synthetic extracellular matrix hydrogels.
Biomacromolecules (2007-08-19)
Monica A Serban, Glenn D Prestwich
ABSTRACT
Hyaluronan (HA) derivatives containing thiol-reactive electrophilic esters were prepared to react with thiol-modified macromolecules to give cross-linker-free hydrogels. Specifically, HA was converted to two haloacetate derivatives, HA bromoacetate (HABA) and HA iodoacetate (HAIA). In cytotoxicity assays, these reactive macromolecules predictably induced cell death in a dose-dependent manner. Cross-linker-free synthetic extracellular matrix (sECM) hydrogels were prepared by thiol alkylation using HAIA and HABA as polyvalent electrophiles and thiol-modified HA (CMHA-S) with or without thiol-modified gelatin (Gtn-DTPH) as polyvalent nucleophiles. When primary human fibroblasts were seeded on the surface of the sECMs containing only the electrophilic HA haloacetate and nucleophilic CMHA-S components, no significant cytoadherence was observed. Cell attachment and viability was 17% (HABA) to 30% (HAIA) lower on HA haloacetate cross-linked hydrogels than on CMHA-S that had been oxidatively cross-linked via disulfide-bonds. In contrast, sECMs that included Gtn-DTPH allowed fibroblasts to attach, spread, and proliferate. Taken together, the HA haloacetates are attractive candidates for producing cross-linker-free sECM biomaterials that can function either as anti-adhesive barriers or as cytoadhesive sECMs for cell culture in pseudo-3-D.
Sigma-Aldrich
S-Acetylmercaptosuccinic anhydride, 96%