MilliporeSigma
  • Amino benzoic acid modified silica--an improved catalyst for the mono-substituted product in the benzylation of toluene with benzyl chloride.
Amino benzoic acid modified silica--an improved catalyst for the mono-substituted product in the benzylation of toluene with benzyl chloride.
Journal of colloid and interface science (2007-03-30)
Farook Adam, Jeyashelly Andas
ABSTRACT
Iron and 4-(methylamino)benzoic acid have been successfully incorporated into silica extracted from rice husk. The silica/Fe/amine complex, RH-Fe(5% amine), showed a ca. 24% increase in specific surface area compared to RH-Fe. This increase was attributed to the templated formation of regular pores. The XRD showed the RH-Fe(5% amine) to be amorphous. The Friedel-Crafts benzylation reaction with toluene using RH-Fe(5% amine) showed a drastic reduction in the di-substituted products to ca. 1.0%.
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Sigma-Aldrich
4-(Methylamino)benzoic acid, 97%