MilliporeSigma
  • Structural elucidation of a 3-O-methyl-D-galactose-containing neutral polysaccharide from the fruiting bodies of Phellinus igniarius.
Structural elucidation of a 3-O-methyl-D-galactose-containing neutral polysaccharide from the fruiting bodies of Phellinus igniarius.
Carbohydrate research (2007-03-16)
Yan Yang, Jingsong Zhang, Yanfang Liu, Qingjiu Tang, Zigao Zhao, Wenshui Xia
ABSTRACT
PIP60-1, a novel heteropolysaccharide isolated from fruiting bodies of the medicinal fungus, Phellinus igniarius, has a molecular weight of 1.71 x 10(4)Da and is composed of L-fucose, D-glucose, D-mannose, D-galactose and 3-O-Me-D-galactose in a ratio of 1:1:1:2:1. A structural investigation of PIP60-1 carried out using sugar and methylation analyses, combined with (1)H and (13)C NMR spectroscopy, including COSY, TOCSY, NOESY, HSQC and HMBC experiments, established the repeating unit of the polysaccharide as the following: [structure: see text]
Sigma-Aldrich
Methyl α-D-galactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Methyl-β-D-galactopyranoside