MilliporeSigma
  • EPR spectra of Cu(2+) doped [Zn(sac)2(dmen)] and [Zn(sac)2(paen)] single crystals.
EPR spectra of Cu(2+) doped [Zn(sac)2(dmen)] and [Zn(sac)2(paen)] single crystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-02-13)
R Biyik, R Tapramaz, O Z Yeşilel
ABSTRACT
Cu(2+) doped single crystals of [Zn(sac)2(dmen)] (sac: saccharinate, dmen: N,N'-dimethylethylendiamine) and [Zn(sac)2(paen)], (paen: N,N'-bis(3-propylamine)ethylendiamine) complexes have been investigated by electron paramagnetic resonance (EPR) technique. Detailed investigations of the EPR spectra indicate that Cu(2+) ion substitute with Zn(2+) ion and forms tetrahedral complex in [Zn(sac)2(dmen)] and octahedral complex in [Zn(sac)2(paen)] hosts. Principal values of the g and hyperfine tensors are determined and the ground state wave functions of Cu(2+) ions are obtained using EPR parameters.
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 98%
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 85%