MilliporeSigma
Isomeric identification by laser control mass spectrometry.
Journal of mass spectrometry : JMS (2006-12-19)
Johanna M Dela Cruz, Vadim V Lozovoy, Marcos Dantus
ABSTRACT
The influence shaped femtosecond laser pulses have on molecular photofragmentation and ionization, coupled with the intrinsic sensitivity of mass spectrometry, results in a powerful tool for fast, accurate, reproducible and quantitative isomeric identification. Complex phase functions are introduced to enhance differences during the laser-molecule interactions, which depend on geometric structure, resulting in different fragmentation fingerprints. A full account is given on the setup and results leading to a technique that can be used to distinguish between compounds normally indistinguishable by conventional electron ionization mass spectrometry. We demonstrate geometric and structural isomer identification of cis-/trans-3-heptene, cis-/trans-4-methyl-2-pentene, o-/p-cresol and o-/p-xylene. For the positional isomers of xylene we present a complete dataset consisting of 1024 different phases to explore phase complexity. A selection of two phases from that data can then be used to achieve quantitative identification in mixtures of xylene isomers. Finally, we evaluate receiver operational curves obtained from our experimental data to demonstrate the reliability that can be achieved by femtosecond laser control mass spectrometry.
Product Description
Supelco
o-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
o-Cresol, SAJ first grade, ≥97.0%
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%