MilliporeSigma
  • Attenuation of the production of inositol 1,4,5-trisphosphate in the mouse vomeronasal organ by antibodies against the alphaq/11 subfamily of G-proteins.
Attenuation of the production of inositol 1,4,5-trisphosphate in the mouse vomeronasal organ by antibodies against the alphaq/11 subfamily of G-proteins.
Chemical senses (2006-06-08)
Roger N Thompson, Audrey Napier, Kennedy S Wekesa
ABSTRACT
The social and reproductive behaviors of most mammals are modulated by pheromones, which are perceived by the vomeronasal organ (VNO). Vomeronasal transduction in vertebrates is activated through G-protein-coupled receptors, which in turn leads to the generation of inositol 1,4,5-trisphosphate (IP(3)) and diacylglycerol (DAG) by the activity of phospholipase C. DAG has been shown to gate the transient receptor potential channel 2, whereas IP(3) may play a role in stimulating the release of calcium from the endoplasmic reticulum store. To investigate the role of the alpha subunits of G(q/11) in the transduction process, microvillar membranes from female mice VNO were preincubated with a selective C-terminal peptide antibody against Galpha(q/11) and then stimulated with adult male urine. Incubation of VNO membranes with antibodies against Galpha(q/11) blocked the production of IP(3) in a dose-dependent manner. We were also able to impair the production of IP(3) when we stimulated with 2-heptanone or 2,5-dimethylpyrazine in the presence of antibodies against the alpha subunit of G(q/11). 2-Heptanone is a known pheromone that has been linked to VIR receptors. Thus, our observations indicate that the alpha subunits of G(q/11) play a role in pheromonal signaling in the VNO.
Sigma-Aldrich
2,5-Dimethylpyrazine, ≥98%, FG
Sigma-Aldrich
2,5-Dimethylpyrazine, 98%