MilliporeSigma
  • Effects of cyanobacterium Fischerella ambigua isolates and cell free culture media on zebrafish (Danio rerio) embryo development.
Effects of cyanobacterium Fischerella ambigua isolates and cell free culture media on zebrafish (Danio rerio) embryo development.
Chemosphere (2006-03-24)
Anthony D Wright, Olaf Papendorf, Gabriele M König, Axel Oberemm
ABSTRACT
The toxic effects of several species of fresh water cyanobacteria, notably Microcystis species and associated toxins, the microcystins, Anabaena species (anatoxin), Nodularia sp. (nodularin), and Cylindrospermopsis raciborskii (cylindrospermopsin), are well known. Little, however, is known about the effects of secondary metabolites other than alkaloids. Early life stage tests with zebrafish (Danio rerio) were used to detect bioactive properties of compounds released by healthy cyanobacteria (Fischerella ambigua), particularly on the early developmental stages of fish. This approach, using F. ambigua is probably most valuable as it shows the toxicity of healthy growing cyanobacteria. The effects of cyanobacterial secondary metabolites on the embryonic stages of fish are of considerable interest as many aquatic creatures, particularly fish, are unable to avoid the potential toxins that may be released by undesirable algal blooms or as a result of allelopathic effects. In the current study, the zebrafish (D. rerio) was used as a model experimental system to investigate the effects of ambigols A and C, tjipanazole D and C, 2,4-dichlorobenzoic acid, cell free culture media, and media extracts of a terrestrial/fresh water strain of the cyanobacterium F. ambigua on embryo development. Fish embryo tests performed with the cell free culture medium showed that after 3h of exposure to undiluted culture medium all fish embryos died. At a tenfold dilution the process of epiboly (formation of the gastrula) was retarded in all embryos, lesions were observed, and their general development was significantly arrested, finally followed by death. The same tests performed with extracts (dichloromethane, n-butanol, and residual cell free culture medium) of the cell free culture medium, ambigol A, ambigol C, 2,4-dichlorobenzoic acid and tjipanazole D showed only ambigol A to have an influence on zebrafish development at concentrations>or=1 mg/l (2.06 microM). After 55 h all embryos showed pectoral oedema, irregularly shaped fin folds, bent tails, and unusual circular neoplasms in the dorsal tail fin fold. Due to the high concentration of ambigol A used in this assay these effects were considered to be of minor importance when compared to those of the culture medium.
Product Description
Supelco
2,4-Dichlorobenzoic acid, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dichlorobenzoic acid, 98%