MilliporeSigma
  • C-terminal N-alkylated peptide amides resulting from the linker decomposition of the Rink amide resin: a new cleavage mixture prevents their formation.
C-terminal N-alkylated peptide amides resulting from the linker decomposition of the Rink amide resin: a new cleavage mixture prevents their formation.
Journal of peptide science : an official publication of the European Peptide Society (2005-08-17)
Panagiotis Stathopoulos, Serafim Papas, Vassilios Tsikaris
ABSTRACT
Decomposition of the resin linkers during TFA cleavage of the peptides in the Fmoc strategy leads to alkylation of sensitive amino acids. The C-terminal amide alkylation, reported for the first time, is shown to be a major problem in peptide amides synthesized on the Rink amide resin. This side reaction occurs as a result of the Rink amide linker decomposition under TFA treatment of the peptide resin. The use of 1,3-dimethoxybenzene in a cleavage cocktail prevents almost quantitatively formation of C-terminal N-alkylated peptide amides. Oxidized by-product in the tested Cys- and Met-containing peptides were not observed, even if thiols were not used in the cleavage mixture.
Sigma-Aldrich
1,3-Dimethoxybenzene, ≥98%, FG
Sigma-Aldrich
1,3-Dimethoxybenzene, ≥98%