MilliporeSigma
  • Ultrafast x-ray diffraction of transient molecular structures in solution.
Ultrafast x-ray diffraction of transient molecular structures in solution.
Science (New York, N.Y.) (2005-07-16)
H Ihee, M Lorenc, T K Kim, Q Y Kong, M Cammarata, J H Lee, S Bratos, M Wulff
ABSTRACT
We report direct structural evidence of the bridged radical (CH2ICH2.) in a polar solution, obtained using time-resolved liquid-phase x-ray diffraction. This transient intermediate has long been hypothesized to explain stereo-chemical control in many association and/or dissociation reactions involving haloalkanes. Ultrashort optical pulses were used to dissociate an iodine atom from the haloethane molecule (C2H4I2) dissolved in methanol, and the diffraction of picosecond x-ray pulses from a synchrotron supports the following structural dynamics, with approximately 0.01 angstrom spatial resolution and approximately 100 picosecond time resolution: The loss of one iodine atom from C2H4I2 leads to the C-I-C triangular geometry of CH2ICH2.. This transient C2H4I then binds to an iodine atom to form a new species, the C2H4I-I isomer, which eventually decays into C2H4 + I2. Solvent dynamics were also extracted from the data, revealing a change in the solvent cage geometry, heating, and thermal expansion.
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
Sigma-Aldrich
Diiodomethane, SAJ first grade, ≥98.0%