MilliporeSigma
  • Enantioselective epoxidation of alpha,beta-enones promoted by alpha,alpha-diphenyl-L-prolinol as bifunctional organocatalyst.
Enantioselective epoxidation of alpha,beta-enones promoted by alpha,alpha-diphenyl-L-prolinol as bifunctional organocatalyst.
Organic letters (2005-06-17)
Alessandra Lattanzi
ABSTRACT
[reaction: see text] An operationally simple and mild protocol for the catalytic enantioselective epoxidation of alpha,beta-unsaturated ketones has been estabilished using commercially available alpha,alpha-diphenyl-l-prolinol as bifunctional organocatalyst and tert-butyl hydroperoxide (TBHP) as oxidant. The epoxides have been obtained in good yields and with up to 80% ee.
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%
Sigma-Aldrich
(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol, 99%