MilliporeSigma
  • Synthesis and insecticidal evaluation of propesticides of benzoylphenylureas.
Synthesis and insecticidal evaluation of propesticides of benzoylphenylureas.
Journal of agricultural and food chemistry (2005-01-06)
Li Chen, Qingmin Wang, Runqiu Huang, Chunhui Mao, Jian Shang, Fuchun Bi
ABSTRACT
Two series of benzoylphenylurea derivatives were synthesized as candidate propesticides by a nucleophilic addition reaction between 2,6-difluronbenzoyl isocyanate and N-substitutedaniline. The new compounds were identified by 1H NMR spectroscopy, electron ionization-mass spectrometry, and elemental analyses. The bioactivities of the new compounds were evaluated. All of the propesticides reported here were soluble in most organic solvents, and their hydrophobicities were improved obviously. The result of the bioactivities of the new compounds against Oriental armyworm showed that some of the new compounds are good as compared to diflubenzuron and penfluron.
Sigma-Aldrich
2,6-Difluorobenzoic acid, 98%