MilliporeSigma
  • Millisecond pulsed radio frequency glow discharge time of flight mass spectrometry: temporal and spatial variations in molecular energetics.
Millisecond pulsed radio frequency glow discharge time of flight mass spectrometry: temporal and spatial variations in molecular energetics.
Journal of the American Society for Mass Spectrometry (2003-12-31)
Lei Li, John T Millay, John P Turner, Fred L King
ABSTRACT
The internal energy distributions, P(epsilon), of a millisecond pulsed radio frequency glow discharge plasma were investigated using tungsten hexcarbonyl W(CO)(6) as a "thermometer molecule". Vapor of the probe molecule, W(CO)(6), was introduced into the plasma and subjected to various ionization and excitation processes therein. The resultant molecular and fragment ions were monitored using a Time-of-Flight mass spectrometer. Ion abundance data were utilized in combination with the known energetics of W(CO)(6) to construct the P(epsilon) plots. The P(epsilon) of W(CO)(6) exhibited strong temporal dependence over the pulse cycle: Distinct internal energy distributions were found at the discharge breakdown period (prepeak), the steady state period (plateau), and the post-pulse period (afterpeak). Spatial variation in P(epsilon) was also observed, especially during the plateau regime. The observations suggest that this pulsed glow discharge affords excellent energy tunability that can be used to perform selective ionization and fragmentation for molecular, structural, and elemental information. Parametric studies were performed to evaluate the effects of discharge pressure and operating power on P(epsilon). These studies also provided insight into the correlation of the observed P(epsilon)s with the fundamental ionization and excitation mechanisms in the plasma. The temporal and spatial variations in P(epsilon) were hence attributed to changes in the dominant energy transfer processes at specific times in specific regions of the plasma. These data will be useful in future efforts to optimize the analytical performance of this source for chemical speciation.
Sigma-Aldrich
Tungsten hexacarbonyl, 97%
Sigma-Aldrich
Tungsten hexacarbonyl, 99.99% trace metals basis (excluding Mo), purified by sublimation
Sigma-Aldrich
Tungsten hexacarbonyl, packaged for use in deposition systems