MilliporeSigma
Toxicological evaluation of dry ice expanded tobacco.
Toxicology letters (2003-10-29)
Eugenia H Theophilus, Dale B Poindexter, Daniel R Meckley, Betsy R Bombick, Michael F Borgerding, Mark A Higuchi, Paul H Ayres, Michael J Morton, Arnold T Mosberg, James E Swauger
ABSTRACT
A tiered testing strategy has been developed to evaluate the potential of tobacco processes, ingredients, or technological developments to change the biological activity resulting from burning tobacco. The strategy is based on comparative chemical and biological testing. Dry ice expanded tobacco (DIET) is an example of a common tobacco expansion process currently used in the manufacture of cigarettes to increase tobacco filling capacity. As part of the toxicological evaluation of DIET, test cigarettes containing DIET were compared with control cigarettes containing tobacco expanded with a traditional expansion agent (Freon-11, also known as trichlorofluoromethane). Testing included mainstream cigarette smoke chemistry studies, genotoxicity studies (Ames and sister chromatid exchange, SCE), a 13-week inhalation study in Sprague-Dawley rats, and a 30-week dermal tumor promotion study in SENCAR mice. Cigarettes containing DIET or Freon-11 expanded tobacco were similar in biological activity.
Sigma-Aldrich
Trichlorofluoromethane solution, NMR reference standard, 10% in acetone-d6 (99.9 atom % D), TMS 6 %, 1,2-dichloro-1,1-difluoroethane 1 %
Sigma-Aldrich
Trichlorofluoromethane, ≥99.5%
Supelco
Trichlorofluoromethane, analytical standard