MilliporeSigma
Cytotoxic effects of antimony trichloride on mice in vivo.
Cytobios (1992-01-01)
N Gurnani, A Sharma, G Talukder
ABSTRACT
Clastogenic effects of antimony trichloride, used in small industries, were monitored in laboratory bred white Swiss mice in vivo, following oral administration by gavaging, after 6, 12, 18 and 24 h. Chromosomal aberrations were principally breaks and damaged cells observed from bone marrow preparations. The frequencies of chromosomal aberrations were directly related to the dose used and were significantly higher than the control. The chemical did not alter the frequency of dividing cells to any significant level.
Sigma-Aldrich
Antimony standard solution, 1000 ppm Sb, suitable for atomic absorption spectrometry
Sigma-Aldrich
Antimony(III) chloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Antimony(III) chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Antimony(III) chloride, ReagentPlus®, 99%