MilliporeSigma
  • [A study on the concentration quenching of Tm3+ upconversion luminescence].
[A study on the concentration quenching of Tm3+ upconversion luminescence].
Guang pu xue yu guang pu fen xi = Guang pu (2003-09-02)
B Chen, H Wang, S Huang
ABSTRACT
In this work, we have a designation and preparation of MFT glasses for upconversion, the glasses consisted of TeO2 and fluoride: PbF2, AlF3, BaF2, NaF and the impurity Tm2O3. In this glass system the oxide improve forming ability, the fluorides improve the microscopic environment around RE ions in glasses. In this glass host the content of Tm2O3 achieves to 4% mol and crystallization no occurred. A detail study on the concentration quenching of upconversion luminescence for 1G4-->3H6 and 1D2-->3H4 transitions was completed. The experimental results directed that the quenching concentration was 0.6 mol.% and higher 3 times than in other glasses materials. The cross relaxation and mechanism of concentration quenching were discussed.
Sigma-Aldrich
Tellurium dioxide, ≥97.0%
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Thulium(III) oxide, 99.9% trace metals basis
Sigma-Aldrich
Thulium(III) oxide, 99.99% trace metals basis