MilliporeSigma
  • [Effect of nicotinamide adenine dinucleotide on the serotoninergic mediator system in the rat brain during 3-acetylpyridine administration].
[Effect of nicotinamide adenine dinucleotide on the serotoninergic mediator system in the rat brain during 3-acetylpyridine administration].
Voprosy meditsinskoi khimii (2002-09-24)
T M Kuchmerovskaia, G V Chichkovskaia, L V Pakirbaeva, A A Khalmuradov
ABSTRACT
The effect of administration of 3-acetylpyridine (antivitamin B3), on rat brain serotoninergic and NAD receptor systems was investigated. The injection of 3-acetylpyridine to rats caused a decrease of NAD and serotonin content in the brain and changes in [U-14C]NAD binding to synaptic membranes, serotonin uptake by nerve endings, as well as sensitivity of this process to NAD (10-5 M). Pretreatment of animals with nicotinamide restored modulating effect of NAD.
Sigma-Aldrich
3-Acetylpyridine, ≥98%, FG
Sigma-Aldrich
3-Acetylpyridine, 98%