MilliporeSigma
Antioxidant properties of 8.0.4'-neolignans.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2002-02-05)
K Kónya, Z S Varga, S Antus
ABSTRACT
A series of naturally occurring 8.0.4'-neolignans (1a-d, 1g, 2g, 2h) and their analogues (le-f, lh, 1i, 2a-f, 2i) have been synthesized in racemic form starting from commercially available phenols, such as eugenol, isoeugenol and 4-allyl-2,6-dimethoxyphenol and from aromatic aldehydes, such as piperonal, veratraldehyde and 3,4,5-trimethoxybenzaldehyde. The inhibitory activity of these compounds on superoxide anion (O2.-) release by human polymorphonuclear leukocytes (PMNLs) was tested and the structure-activity relationship was also studied.
Sigma-Aldrich
4-Allyl-2,6-dimethoxyphenol, ≥95%
Sigma-Aldrich
4-Allyl-2,6-dimethoxyphenol, ≥95%, FG
Sigma-Aldrich
3,4-Dimethoxybenzaldehyde, 99%
Sigma-Aldrich
Veratraldehyde, ≥98%, FG
Sigma-Aldrich
Piperonal, ≥99%, FCC, FG
Sigma-Aldrich
Piperonal, 99%