MilliporeSigma
Conversion of methionine to methional by Lactococcus lactis.
FEMS microbiology letters (2001-10-30)
F Amárita, D Fernández-Esplá, T Requena, C Pelaez
ABSTRACT
Lactic acid bacteria were screened for methional production from 4-methylthio-2-ketobutanoate. Only Lactococcus lactis IFPL730 produced high amounts of methional. It was demonstrated that production of this compound was an exclusively enzymatic reaction. The present work describes for the first time that L. lactis can convert enzymatically methionine to methional in a process mediated by aminotransferase and alpha-ketoacid decarboxylase activities. The activity seems to be strain dependent.
Sigma-Aldrich
3-(Methylthio)propionaldehyde, 96%
Sigma-Aldrich
Methional, ≥97%, FG
Sigma-Aldrich
Methional, natural, ≥98%, FG