MilliporeSigma
  • Concise synthesis of ciguatoxin ABC-ring fragments and surface plasmon resonance study of the interaction of their BSA conjugates with monoclonal antibodies.
Concise synthesis of ciguatoxin ABC-ring fragments and surface plasmon resonance study of the interaction of their BSA conjugates with monoclonal antibodies.
Bioorganic & medicinal chemistry letters (2001-07-17)
Y Nagumo, H Oguri, Y Shindo, S Sasaki, T Oishi, M Hirama, Y Tomioka, M Mizugaki, T Tsumuraya
ABSTRACT
Monoclonal antibodies (mAbs), 4H2 and 6H7, were prepared previously using a protein conjugate of a 1:1 epimeric mixture of the synthetic ABC-ring fragments of ciguatoxin (CTX), 3 and 4. Here, the interactions of these mAbs with the fragments of CTX and CTX3C, 3 and 5, were investigated by surface plasmon resonance (SPR) spectroscopy in an attempt to clarify an antigenic determinant. Compared with the previous synthesis, the fragment 3 possessing the 2S configuration was synthesized from tri-O-acetyl-D-glucal much more effectively. The mAb 4H2 was already known to show a dose-dependent binding to the bovine serum albumin (BSA) conjugate of 3, but not to that of 5. The present SPR study of 4H2 demonstrates that the A-ring side chain of 3 plays a decisive role as an epitope. Therefore, SPR can effectively replace the ELISA method for the analysis of mAbs.
Sigma-Aldrich
D-Glucal, 96%