MilliporeSigma
Novel biomarkers of the metabolism of caffeic acid derivatives in vivo.
Free radical biology & medicine (2001-05-23)
A R Rechner, J P Spencer, G Kuhnle, U Hahn, C A Rice-Evans
ABSTRACT
The purpose of this study was to investigate biomarkers of the bioavailability and metabolism of hydroxycinnamate derivatives through the determination of the pharmacokinetics of their urinary elimination and identification of the metabolites excreted. Coffee was used as a rich source of caffeic acid derivatives and human supplementation was undertaken. The results show a highly significant increase in the excretion of ferulic, isoferulic, dihydroferulic acid (3-(4-hydroxy-3-methoxyphenyl)-propionic acid), and vanillic acid postsupplementation relative to the levels presupplementation. Thus, ferulic, isoferulic, and dihydroferulic acids are specific biomarkers for the bioavailability and metabolism of dietary caffeic acid esters. Isoferulic acid is a unique biomarker as it is not a dietary component, however, dihydroferulic acid may well derive from other flavonoids with a structurally related B-ring. 3-Hydroxyhippuric acid has also been identified as an indicator for bioavailability and metabolism of phenolic compounds, and shows a highly significant excretion increase postsupplementation. The results reveal isoferulic acid (and possibly dihydroferulic acid) as novel markers of caffeoyl quinic acid metabolism.
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%