MilliporeSigma
  • Colorimetric determination of tocopheryl acetate (vitamin E) in pure form and in multi-vitamin capsules.
Colorimetric determination of tocopheryl acetate (vitamin E) in pure form and in multi-vitamin capsules.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2001-05-10)
A S Amin
ABSTRACT
A simple, accurate and sensitive method for the determination of micro amounts of vitamin E (V(E)) in pure form and in multi-vitamin capsules is described. The method is based on the reduction of tetrazolium blue in slightly alkaline medium by vitamin E after extraction from aqueous EDTA medium with petroleum ether and transesterification. The oxidation reduction reaction occurs after 10 min of heating in a water bath at 90+/-2 degrees C, leading to the formation of a highly coloured formazan derivative. Different variables affecting the colour development were investigated and optimized. Absorbance measurements were made at 526 nm and the calibration graph was linear for 0.2-11.0 microg/ml of (V(E)) with relative precision of about 0.7-1.5% when the standard additions method is used.
Sigma-Aldrich
Tetrazolium Blue Chloride, suitable for microbiology, ≥90% (T)
Sigma-Aldrich
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
Supelco
Tocopheryl acetate (Vitamin E acetate) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®