MilliporeSigma
  • Enantiomer separation of alpha-ionone using gas chromatography with cyclodextrin derivatives as chiral stationary phases.
Enantiomer separation of alpha-ionone using gas chromatography with cyclodextrin derivatives as chiral stationary phases.
Journal of chromatography. A (2000-02-16)
F Quattrini, G Biressi, M Juza, M Mazzotti, C Fuganti, M Morbidelli
ABSTRACT
The gas chromatographic enantiomer separation of alpha-ionone was studied with three different chiral stationary phases using as chiral selectors: (1) heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin, dissolved in polysiloxane PS-086, (2) octakis(2,6-di-O-pentyl-3-O-trifluoroacetyl)-gamma-cyclodextrin and (3) octakis(2,6-di-O-pentyl-3-O-butanoyl)-gamma-cyclodextrin, both dissolved in polysiloxane SE-54. The influence of the concentration of the chiral selector in the polysiloxane, coated on Chromosorb P AW-DMCS 80-100 mesh, is described and discussed, as well as the effect of Chromosorb loading. The feasibility of the preparative gas chromatographic separation of the enantiomers of alpha-ionone is considered; in order to provide a term of comparison, the estimated performances are compared with those achieved in the separation of the enantiomers of the inhalation anaesthetic enflurane.
Sigma-Aldrich
α-Ionone, ≥90%, stabilized
Sigma-Aldrich
α-Ionone, technical grade, 90%