MilliporeSigma
  • Small molecule inhibition of phosphatidylinositol-3,4,5-triphosphate (PIP3) binding to pleckstrin homology domains.
Small molecule inhibition of phosphatidylinositol-3,4,5-triphosphate (PIP3) binding to pleckstrin homology domains.
Proceedings of the National Academy of Sciences of the United States of America (2010-11-03)
Benchun Miao, Igor Skidan, Jinsheng Yang, Alexey Lugovskoy, Mikhail Reibarkh, Kai Long, Tres Brazell, Kulbhushan A Durugkar, Jenny Maki, C V Ramana, Brian Schaffhausen, Gerhard Wagner, Vladimir Torchilin, Junying Yuan, Alexei Degterev
ABSTRACT
The PI3-kinase (PI3K) pathway regulates many cellular processes, especially cell metabolism, cell survival, and apoptosis. Phosphatidylinositol-3,4,5-trisphosphate (PIP3), the product of PI3K activity and a key signaling molecule, acts by recruiting pleckstrin-homology (PH) domain-containing proteins to cell membranes. Here, we describe a new structural class of nonphosphoinositide small molecule antagonists (PITenins, PITs) of PIP3-PH domain interactions (IC(50) ranges from 13.4 to 31 μM in PIP3/Akt PH domain binding assay). PITs inhibit interactions of a number of PIP3-binding PH domains, including those of Akt and PDK1, without affecting several PIP2-selective PH domains. As a result, PITs suppress the PI3K-PDK1-Akt pathway and trigger metabolic stress and apoptosis. A PIT-1 analog displayed significant antitumor activity in vivo, including inhibition of tumor growth and induction of apoptosis. Overall, our studies demonstrate the feasibility of developing specific small molecule antagonists of PIP3 signaling.
Sigma-Aldrich
Pit-1, ≥98% (HPLC)
Sigma-Aldrich
PIP3 Antagonist II, DM-PIT-1, The PIP3 Antagonist II, DM-PIT-1, also referenced under CAS 701947-53-7, controls the biological activity of PIP3.