MilliporeSigma
  • High-sensitivity fluorescence lifetime thermal sensing based on CdTe quantum dots.
High-sensitivity fluorescence lifetime thermal sensing based on CdTe quantum dots.
Small (Weinheim an der Bergstrasse, Germany) (2012-06-16)
P Haro-González, L Martínez-Maestro, I R Martín, J García-Solé, D Jaque
ABSTRACT
The potential use of CdTe quantum dots as luminescence nano-probes for lifetime fluorescence nano-thermometry is demonstrated. The maximum thermal sensitivity achievable is strongly dependent on the quantum dot size. For the smallest sizes (close to 1 nm) the lifetime thermal sensitivity overcomes those of conventional nano-probes used in fluorescence lifetime thermometry.
Sigma-Aldrich
CdTe core-type quantum dots, COOH functionalized, fluorescence λem 710 nm, powder
Sigma-Aldrich
CdTe core-type quantum dots, COOH functionalized, fluorescence λem 770 nm, powder
Sigma-Aldrich
CdTe core-type quantum dots, COOH functionalized, fluorescence λem 520 nm, powder
Sigma-Aldrich
CdTe core-type quantum dots, COOH functionalized, fluorescence λem 570 nm, powder
Sigma-Aldrich
CdTe core-type quantum dots, COOH functionalized, fluorescence λem 610 nm, powder