MilliporeSigma
  • Treatment with BX471, a CC chemokine receptor 1 antagonist, attenuates systemic inflammatory response during sepsis.
Treatment with BX471, a CC chemokine receptor 1 antagonist, attenuates systemic inflammatory response during sepsis.
American journal of physiology. Gastrointestinal and liver physiology (2007-01-20)
Min He, Richard Horuk, Shabbir M Moochhala, Madhav Bhatia
ABSTRACT
Sepsis is a complex clinical syndrome resulting from a harmful host inflammatory response to infection. Chemokines and their receptors play a key role in the pathogenesis of sepsis. BX471 is a potent nonpeptide CC chemokine receptor-1 (CCR1) antagonist in both human and mouse. The aim of the present study was to evaluate the effect of prophylactic and therapeutic treatment with BX471 on cecal ligation and puncture-induced sepsis in the mouse and to investigate the underlying mechanisms. In sepsis induced by cecal ligation and puncture, treatment with BX471 significantly protected mice against lung and liver injury by attenuating MPO activity, an indicator of neutrophil recruitment in lungs and livers and attenuating lung and liver morphological changes in histological sections. Blocking CCR1 by BX471 also downregulated ICAM-1, P-selectin, and E-selectin expression at mRNA and protein levels in lungs and livers compared with placebo-treated groups. These findings suggest that blockage of CCR1 by specific antagonist may represent a promising strategy to prevent disease progression in sepsis.
Sigma-Aldrich
BX471, ≥98% (HPLC)