MilliporeSigma
Enhancing results of microarray hybridizations through microagitation.
Journal of biomolecular techniques : JBT (2003-09-19)
Andreas Toegl, Roland Kirchner, Christoph Gauer, Achim Wixforth
ABSTRACT
Protein and DNA microarrays have become a standard tool in proteomics/genomics research. In order to guarantee fast and reproducible hybridization results, the diffusion limit must be overcome. Surface acoustic wave (SAW) micro-agitation chips efficiently agitate the smallest sample volumes (down to 10 microL and below) without introducing any dead volume. The advantages are reduced reaction time, increased signal-to-noise ratio, improved homogeneity across the microarray, and better slide-to-slide reproducibility. The SAW micromixer chips are the heart of the Advalytix Array-Booster, which is compatible with all microarrays based on the microscope slide format.
Sigma-Aldrich
Denhardt′s Solution 50x, for Northern and Southern blotting, solution
Sigma-Aldrich
Denhardt′s Solution, for Northern and Southern blotting, lyophilized powder