MilliporeSigma
  • Carbonylative Heck reactions using CO generated ex situ in a two-chamber system.
Carbonylative Heck reactions using CO generated ex situ in a two-chamber system.
Organic letters (2011-04-08)
Philippe Hermange, Thomas M Gøgsig, Anders T Lindhardt, Rolf H Taaning, Troels Skrydstrup
ABSTRACT
A carbonylative Heck reaction of aryl iodides and styrene derivatives employing a two-chamber system using a stable, crystalline, and nontransition metal based carbon monoxide source is reported. By applying near-stoichiometric amounts of the carbon monoxide precursor, an effective exploitation of the hazardous CO gas is obtained affording chalcone derivatives in good yields. Application to isotope labeling, incorporating (13)CO, was further established.
Sigma-Aldrich
9-Methyl-9H-fluorene-9-carbonyl chloride, ≥99.0% (GC)
Sigma-Aldrich
9-Methyl-9H-fluorene-9-carbonyl-13C chloride, ≥97.0% (GC)