MilliporeSigma
  • Isolation and characterization of an endosperm-specific promoter from wheat (Triticum aestivum L.).
Isolation and characterization of an endosperm-specific promoter from wheat (Triticum aestivum L.).
Zeitschrift fur Naturforschung. C, Journal of biosciences (2013-02-19)
Fei Song, Cui-Ju Cui, Ling Chen, Yang-Liu Sun, Fei-Fei Wang, Javeed Hussain, Yin Li, Chen Wang, Cheng Wang, Ming-Jie Chen, Yue-Sheng Wang, Guang-Xiao Yang, Guang-Yuan He
ABSTRACT
Genes coding for avenin-like proteins (ALP) represent a new family of wheat storage protein genes. To find a wheat endosperm-specific promoter, a 1644-bp fragment upstream of the ALP type-B gene (GenBank accession number JN622144) was isolated. The important promoter elements of the ALP type-B gene were ascertained through sequence analysis which revealed that this fragment contains the TATA and CAAT boxes, which are important elements in gene expression. A prolamin box containing an endosperm motif and a GCN4-like motif (GLM) is present at about 300 bp upstream of the translation start site. The promoter sequence has two ESP-like elements and one of them is followed by an RY motif with the nucleotides CATG overlapping. The RY motif is considered the core functional sequence in a promoter. In an attempt to confirm the promoter activity, a series of 5'-deletions of the promoter were fused with the beta-glucuronidase (GUS) gene, and the constructs were stably introduced into tobacco plants. GUS staining confirmed that the AVL type-B promoter is an endosperm-specific promoter in tobacco seeds. Quantitative analysis of GUS expression in transgenic plants showed that even the shortest 5'-deletion, i.e. a 290-bp promoter sequence within the prolamin box, was sufficient to drive GUS expression in the endosperm. The highest expression level was found in transgenic plants containing the 5'-deletion vector construct pALP-8. This suggests that the ESP-like element overlapping with the RY motif may play a crucial role in the regulatory function of the promoter.
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid