MilliporeSigma
  • Purification, biochemical characterization and dye decolorization capacity of an alkali-resistant and metal-tolerant laccase from Trametes pubescens.
Purification, biochemical characterization and dye decolorization capacity of an alkali-resistant and metal-tolerant laccase from Trametes pubescens.
Bioresource technology (2012-12-01)
Jing Si, Feng Peng, Baokai Cui
ABSTRACT
Extracellular laccase (Tplac) from Trametes pubescens was purified to homogeneity by a three-step method, which resulted in a high specific activity of 18.543 Umg(-1), 16.016-fold greater than that of crude enzyme at the same level. Tplac is a monomeric protein that has a molecular mass of 68 kDa. The enzyme demonstrated high activity toward 1.0mM ABTS at an optimum pH of 5.0 and temperature of 50 °C, and under these conditions, the catalytic efficiency (k(cat)/K(m)) is 8.34 s(-1) μM(-1). Tplac is highly stable and resistant under alkaline conditions, with pH values ranging from 7.0 to 10.0. Interestingly, above 88% of initial enzyme activity was maintained in the presence of metal ions at 25.0mM, leading to an increase in substrate affinity, which indicated that the laccase is highly metal-tolerant. These unusual properties demonstrated that the new fungal laccase Tplac has potentials for the specific industrial or environmental applications.
Sigma-Aldrich
Laccase from Rhus vernicifera, crude acetone powder, ≥50 units/mg solid
Sigma-Aldrich
Laccase from Trametes versicolor, powder, light brown, ≥0.5 U/mg
Sigma-Aldrich
Laccase from Agaricus bisporus, powder, deep brown, ≥4 U/mg