MilliporeSigma
  • Crystallization and preliminary X-ray diffraction analysis of an L-amino-acid oxidase from Bothrops jararacussu venom.
Crystallization and preliminary X-ray diffraction analysis of an L-amino-acid oxidase from Bothrops jararacussu venom.
Acta crystallographica. Section F, Structural biology and crystallization communications (2012-02-03)
Anwar Ullah, Monika Coronado, Mário T Murakami, Christian Betzel, Raghuvir K Arni
ABSTRACT
Snake-venom L-amino-acid oxidases (SV-LAAOs) trigger a wide range of local and systematic effects, including inhibition of platelet aggregation, cytotoxicity, haemolysis, apoptosis and haemorrhage. These effects mainly arise from the uncontrolled release of the hydrogen peroxide that is produced by the redox reaction involving L-amino acids catalyzed by these flavoenzymes. Taking their clinical relevance into account, few SV-LAAOs have been structurally characterized and the structural determinants responsible for their broad direct and indirect pharmacological activities remain unclear. In this work, an LAAO from Bothrops jararacussu venom (BJu-LAAO) was purified and crystallized. The BJu-LAAO crystals belonged to space group P2(1), with unit-cell parameters a = 66.38, b = 72.19, c = 101.53 Å, β = 90.9°. The asymmetric unit contained two molecules and the structure was determined and partially refined at 3.0 Å resolution.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥4.0 units/mg protein, aqueous suspension
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type I (dried venom)