MilliporeSigma
  • [Trichoderma produces an inhibitor of the human immunodeficiency virus].
[Trichoderma produces an inhibitor of the human immunodeficiency virus].
Voprosy meditsinskoi khimii (2000-11-15)
I P Smirnova, S B Alekseev, D A Moshkov
ABSTRACT
The ability of protein isolated from (Trichoderma Rifai) and azydothymidine to inhibit the reproduction of HIV-virus was compared. The obtained experimental data have verified that Trichoderma Rifai protein is a promising human immunodeficiency virus (HIV) inhibitor.
Sigma-Aldrich
Lysine Oxidase from Trichoderma viride, lyophilized powder, ≥20 units/mg protein