MilliporeSigma
Microbial production and applications of chiral hydroxyalkanoates.
Applied microbiology and biotechnology (2005-02-09)
Guo-Qiang Chen, Qiong Wu
ABSTRACT
Polyhydroxyalkanoates (PHA) are a family of polyesters consisting of over 150 chiral hydroxyalkanoic acids (HA). This paper reviews the physiological functions of (R)-3-hydroxybutyric acid (3HB) and (R)-4-hydroxybutyric acid and summarizes the technologies developed to produce various HA [3HB, (R)-3-hydroxyoctanoic acid, (R)-3-hydroxydecanoic acid, etc.] and the applications of chiral HA. Their outlooks and perspectives are discussed.
Sigma-Aldrich
Oligo-HA4, low endotoxin
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%