MilliporeSigma
  • Production of biosurfactant lipopeptides Iturin A, fengycin and surfactin A from Bacillus subtilis CMB32 for control of Colletotrichum gloeosporioides.
Production of biosurfactant lipopeptides Iturin A, fengycin and surfactin A from Bacillus subtilis CMB32 for control of Colletotrichum gloeosporioides.
Journal of microbiology and biotechnology (2010-02-06)
Pyoung Il Kim, Jaewon Ryu, Young Hwan Kim, Youn-Tae Chi
ABSTRACT
A bacterial strain isolated from soil for its potential to control the anthracnose disease caused by Colletotrichum gloeosporioides was identified as a Bacillus subtilis. Bacillus subtilis CMB32 produced antifungal agents on M9 broth at 30degreesC. Biosurfactant lipopeptides produced by Bacillus subtilis CMB32 were precipitated by adjusting to pH 2 and extracting using chloroform/methanol, and then were purified using column chromatography and reverse-phase HPLC. Molecular masses of the lipopeptides were estimated by MALDI-TOF mass spectrometry as (a) 1080, (b) 1486, and (c) 1044 Da, respectively. They had cyclic structures and amino acid compositions of (a) Pro, Asx, Ser, Tyr, Glx, (b) Glx, Tyr, Thr, Ala, Pro, Ile, and (c) Glx, Leu, Val, Asx, respectively. Further analysis revealed that Bacillus subtilis CMB32 produced three antifungal lipopeptides: (a) iturin A, (b) fengycin, and (c) surfactin A.
Sigma-Aldrich
Iturin A from Bacillus subtilis, ≥95% (HPLC)
Millipore
M9 Broth, suitable for microbiology, NutriSelect® Basic