MilliporeSigma
  • Biocompatibility of Fe(3)O(4) nanoparticles evaluated by in vitro cytotoxicity assays using normal, glia and breast cancer cells.
Biocompatibility of Fe(3)O(4) nanoparticles evaluated by in vitro cytotoxicity assays using normal, glia and breast cancer cells.
Nanotechnology (2010-01-22)
B Ankamwar, T C Lai, J H Huang, R S Liu, M Hsiao, C H Chen, Y K Hwu
ABSTRACT
In order to reveal the biocompatibility of Fe(3)O(4) nanoparticles and bipolar surfactant tetramethylammonium 11-aminoundecanoate cytotoxicity tests were performed as a function of concentration from low (0.1 microg ml(-1)) to higher concentration (100 microg ml(-1)) using various human glia, human breast cancer and normal cell lines. Cytotoxicity tests for human glia (D54MG, G9T, SF126, U87, U251, U373), human breast cancer (MB157, SKBR3, T47D) and normal (H184B5F5/M10, WI-38, SVGp12) cell lines exhibited almost nontoxicity and reveal biocompatibility of Fe(3)O(4) nanoparticles in the concentration range of 0.1-10 microg ml(-1), while accountable cytotoxicity can be seen at 100 microg ml(-1). The results of our studies suggest that Fe(3)O(4) nanoparticles coated with bipolar surfactant tetramethylammonium 11-aminoundecanoate are biocompatible and promising for bio-applications such as drug delivery, magnetic resonance imaging and magnetic hyperthermia.
Sigma-Aldrich
11-Aminoundecanoic acid, 97%