MilliporeSigma
  • An essential signaling function of cytoplasmic NELFB is independent of RNA polymerase II pausing.
An essential signaling function of cytoplasmic NELFB is independent of RNA polymerase II pausing.
The Journal of biological chemistry (2023-09-18)
Haihui Pan, Xiaolong Cheng, Pedro Felipe Gardeazábal Rodríguez, Xiaowen Zhang, Inhee Chung, Victor X Jin, Wei Li, Yanfen Hu, Rong Li
ABSTRACT
The four-subunit negative elongation factor (NELF) complex mediates RNA polymerase II (Pol II) pausing at promoter-proximal regions. Ablation of individual NELF subunits destabilizes the NELF complex and causes cell lethality, leading to the prevailing concept that NELF-mediated Pol II pausing is essential for cell proliferation. Using separation-of-function mutations, we show here that NELFB function in cell proliferation can be uncoupled from that in Pol II pausing. NELFB mutants sequestered in the cytoplasm and deprived of NELF nuclear function still support cell proliferation and part of the NELFB-dependent transcriptome. Mechanistically, cytoplasmic NELFB physically and functionally interacts with prosurvival signaling kinases, most notably phosphatidylinositol-3-kinase/AKT. Ectopic expression of membrane-tethered phosphatidylinositol-3-kinase/AKT partially bypasses the role of NELFB in cell proliferation, but not Pol II occupancy. Together, these data expand the current understanding of the physiological impact of Pol II pausing and underscore the multiplicity of the biological functions of individual NELF subunits.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-ATP Synthase subunit β Antibody, clone 11/21-7-A8, clone 11/21-7-A8, from mouse
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse